missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Magnesium Nitrate Matrix Modifier for AAS 10,000 ppm in H2O, Ricca Chemical

Catalog No. RV01013050N
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV01013050N 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV01013050N Supplier Ricca Chemical Company Supplier No. RV01013050N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 10,000 ppm in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

Chemical Name or Material Magnesium Nitrate AAS Matrix Modifier
Packaging Natural Poly Bottle
CAS 10377-60-3, 7732-18-5
CAS Min % 97.02%, 0.98%
CAS Max % 100%, 1.02%
Specific Gravity 1.01
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
For Use With (Equipment) Atomic Absorption Spectrophotometer
Solubility Miscible with water
Solution Type Calibration Standard
Type Matrix Modifier
Quantity 50 mL
Grade High Purity
Show More Show Less