accessibility menu, dialog, popup

UserName

CAS RN 10332-33-9

IUPAC Name: sodium hydrate [(oxoboranyl)oxy]olate
Synonyms: sodium hydrate (oxoboranyl)oxyolate
Molecular Weight (g/mol): 99.81
Molecular Formula: BH2NaO4
InChi Key: MDGXUEVTGARGDK-UHFFFAOYSA-M
SMILES: O.[Na+].[O-]OB=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (4)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)