accessibility menu, dialog, popup

UserName

NH2NH2NH2NOHNOO

CAS RN 10308-82-4

IUPAC Name: N''-aminoguanidine; nitric acid
Synonyms: amino-guanidine; nitric acid
Molecular Weight (g/mol): 137.1
Molecular Formula: CH7N5O3
InChi Key: PMGFHEJUUBDCLU-UHFFFAOYSA-N
SMILES: O[N+]([O-])=O.NN=C(N)N
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)