accessibility menu, dialog, popup

UserName

NH2O

CAS RN 103-81-1

IUPAC Name: 2-phenylacetamide
Synonyms: phenylacetamide phenyl-β-acetylamine α-toluamide
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: LSBDFXRDZJMBSC-UHFFFAOYSA-N
SMILES: NC(=O)CC1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)