accessibility menu, dialog, popup

UserName

CH3OO(E)

CAS RN 103-26-4

IUPAC Name: methyl (2E)-3-phenylprop-2-enoate
Synonyms: methyl cinnamate
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: CCRCUPLGCSFEDV-BQYQJAHWSA-N
SMILES: COC(=O)\C=C\C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)