missing translation for 'onlineSavingsMsg'
Learn More

Methyl cinnamate, predominantly trans, 99%

Catalog No. AAA1597536
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1597536 500 g
AAA1597522 100 g
AAA159750E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1597536 Supplier Thermo Scientific Chemicals Supplier No. A1597536
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 103-26-4 | C10H10O2 | 162.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Methyl cinnamate was used to inhibit monophenolase and diphenolase activity of mushroom tyrosinase and it also has antimicrobial ability. It is mainly is used in the flavor and perfume industries. It is known to attract males of various orchid bees, such as Aglae caerulea.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 103-26-4
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00008458
InChI Key CCRCUPLGCSFEDV-BQYQJAHWSA-N
Synonym methyl cinnamate, methyl trans-cinnamate, methyl e-cinnamate, cinnamic acid methyl ester, methyl cinnamylate, methyl 3-phenylacrylate, methyl e-3-phenylprop-2-enoate, methyl 3-phenylpropenoate, trans-cinnamic acid methyl ester, trans-methyl cinnamate
PubChem CID 637520
ChEBI CHEBI:6857
IUPAC Name methyl (E)-3-phenylprop-2-enoate
SMILES COC(=O)C=CC1=CC=CC=C1

Specifications

Melting Point 34°C to 37°C
Density 1.042
Boiling Point 262°C to 264°C
Flash Point 123°C (253°F)
Assay Percent Range 99%
Refractive Index 1.577
Quantity 500 g
Beilstein 386468
Merck Index 14,2299
Solubility Information Insoluble in water.
Formula Weight 162.19
Percent Purity 99%
Chemical Name or Material Methyl cinnamate, predominantly trans
Show More Show Less
EINECSNumber 203-093-8
RTECSNumber GE0190000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only