missing translation for 'onlineSavingsMsg'
Learn More

Methyl cinnamate, 98%

Catalog No. AC279765000
Change view
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC279765000 500 g Glass bottle
AC279760050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC279765000 Supplier Thermo Scientific Chemicals Supplier No. 279765000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 103-26-4 | C10H10O2 | 162.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103-26-4
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00008458
InChI Key CCRCUPLGCSFEDV-BQYQJAHWSA-N
Synonym methyl cinnamate, methyl trans-cinnamate, methyl e-cinnamate, cinnamic acid methyl ester, methyl cinnamylate, methyl 3-phenylacrylate, methyl e-3-phenylprop-2-enoate, methyl 3-phenylpropenoate, trans-cinnamic acid methyl ester, trans-methyl cinnamate
PubChem CID 637520
ChEBI CHEBI:6857
IUPAC Name methyl (E)-3-phenylprop-2-enoate
SMILES COC(=O)C=CC1=CC=CC=C1

Specifications

Acidity 1mg KOH/g max.
Melting Point 33.5°C to 36°C
Color White to Yellow
Boiling Point 260°C to 262°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH=CHCO2CH3
Quantity 500 g
Beilstein 09, 581
Merck Index 15, 2299
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,glycerol,most fixed,oils and mineral oil
Formula Weight 162.19
Percent Purity 98%
Physical Form Fused Crystalline Mass
Chemical Name or Material Methyl cinnamate, 98%
Show More Show Less
EINECSNumber 203-093-8
RTECSNumber GE0190000
TSCA TSCA

RUO – Research Use Only