missing translation for 'onlineSavingsMsg'
Learn More

Methyl Cinnamate 99.0+%, TCI America™

Catalog No. C0360500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0360500G 500 g
C036025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0360500G Supplier TCI America Supplier No. C0360500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 103-26-4
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00008458
InChI Key CCRCUPLGCSFEDV-BQYQJAHWSA-N
Synonym methyl cinnamate, methyl trans-cinnamate, methyl e-cinnamate, cinnamic acid methyl ester, methyl cinnamylate, methyl 3-phenylacrylate, methyl e-3-phenylprop-2-enoate, methyl 3-phenylpropenoate, trans-cinnamic acid methyl ester, trans-methyl cinnamate
PubChem CID 637520
ChEBI CHEBI:6857
IUPAC Name methyl (E)-3-phenylprop-2-enoate
SMILES COC(=O)C=CC1=CC=CC=C1

Specifications

Color White
Boiling Point 262°C
Quantity 500 g
Formula Weight 162.19
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl Cinnamate
EINECSNumber (3)-1750
RTECSNumber GE0190000
TSCA Yes