CAS RN 1027642-30-3
IUPAC Name:
potassium trifluoro(phenoxymethyl)boranuide
Synonyms:
potassium trifluoro(phenoxymethyl)boranuide
Molecular Weight (g/mol):
214.04
Molecular Formula:
C7H7BF3KO
InChi Key:
OVYSNXCRBZPJIG-UHFFFAOYSA-N
SMILES:
[K+].F[B-](F)(F)COC1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
Potassium phenoxymethyltrifluoroborate, 95%, Thermo Scientific Chemicals
Potassium phenoxymethyltrifluoroborate, 95%, Thermo Scientific Chemicals
CAS: 1027642-30-3 Molecular Formula: C7H7BF3KO Molecular Weight (g/mol): 214.036 MDL Number: MFCD10566516 InChI Key: OVYSNXCRBZPJIG-UHFFFAOYSA-N Synonym: potassium phenoxy-methyltrifluoroborate,potassium trifluoro phenoxymethyl boranuide,pubchem11600,potassium phenoxymethyltrifluoroborate,potassium trifluoro phenoxymethyl borate,potassium trifluoro phenoxymethyl borate 1-,potassium phenoxymethyltrifluoroborate, contains kbr PubChem CID: 45479874 IUPAC Name: potassium;trifluoro(phenoxymethyl)boranuide SMILES: [B-](COC1=CC=CC=C1)(F)(F)F.[K+]