accessibility menu, dialog, popup

UserName

CAS RN 102268-15-5

H3CNOHOHOO

CAS RN 102268-15-5

IUPAC Name: 5-ethylpyridine-2,3-dicarboxylic acid
Synonyms: 5-ethylpyridine-2,3-dicarboxylic acid
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: MTAVBTGOXNGCJR-UHFFFAOYSA-N
SMILES: CCC1=CN=C(C(O)=O)C(=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results