accessibility menu, dialog, popup

UserName

OHOOHHO

CAS RN 102-32-9

IUPAC Name: 2-(3,4-dihydroxyphenyl)acetic acid
Synonyms: 3,4 dihydroxyphenylacetic acid homoprotocatechuate dopacetic acid DHY HAA dopacetate dopac dihydroxyphenylacetic acid (substance) 3,4-dhpop homoprotocatechuic acid
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: CFFZDZCDUFSOFZ-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(O)C(O)=C1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)