missing translation for 'onlineSavingsMsg'
Learn More

BES, 99%

Catalog No. AAA1609236
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1609236 500 g
AAA1609214 25 g
AAA1609222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1609236 Supplier Thermo Scientific Chemicals Supplier No. A1609236
Only null left

CAS: 10191-18-1 | C6H15NO5S | 213.25 g/mol

BES is used as a buffer to maintain the pH of the solutions in biological experiments. It is useful for diagnostic assay manufacturing industry. It induces cross-linking between sulfonated polyimide chains in sulfonated polyimide membranes. It acts as biobuffer to investigate the aqueous medium self-assembly of heterometallic CuII/Li 3D coordination polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 10191-18-1
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00007533
InChI Key AJTVSSFTXWNIRG-UHFFFAOYSA-N
Synonym n,n-bis 2-hydroxyethyl-2-aminoethanesulfonic acid, bes, ethanesulfonic acid, 2-bis 2-hydroxyethyl amino, 2-bis 2-hydroxyethyl amino ethanesulfonic acid, n,n-bis 2-hydroxyethyl taurine, n,n-bis hydroxyethyl-2-aminoethanesulfonic acid, bes buffering agent, taurine, n,n-bis 2-hydroxyethyl, 2-bis 2-hydroxyethyl amino-ethanesulfonicacid
PubChem CID 73243
ChEBI CHEBI:39045
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethanesulfonic acid
SMILES OCCN(CCO)CCS(O)(=O)=O

Specifications

Melting Point 153°C to 157°C
Quantity 500 g
Beilstein 1781572
Solubility Information Soluble in water.
Formula Weight 213.25
Percent Purity 99%
Chemical Name or Material BES
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-465-5
RTECSNumber KI7363500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only