accessibility menu, dialog, popup

UserName

ClNClClNNOHNO

CAS RN 101831-37-2

IUPAC Name: 2-(4-chlorophenyl)-2-[2,6-dichloro-4-(3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazin-2-yl)phenyl]acetonitrile
Synonyms: diclazuril
Molecular Weight (g/mol): 407.64
Molecular Formula: C17H9Cl3N4O2
InChi Key: ZSZFUDFOPOMEET-UHFFFAOYNA-N
SMILES: ClC1=CC=C(C=C1)C(C#N)C1=C(Cl)C=C(C=C1Cl)N1N=CC(=O)NC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results