accessibility menu, dialog, popup

UserName

H3CNNN

CAS RN 1014403-09-8

IUPAC Name: 8-(3-methylphenyl)-4,8,12-triazatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Synonyms: 8-(3-methylphenyl)-4,8,12-triazatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular Weight (g/mol): 259.31
Molecular Formula: C17H13N3
InChi Key: WAQDNMLJAYQJRN-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1)N1C2=C(C=NC=C2)C2=C1C=CN=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results