accessibility menu, dialog, popup

UserName

HCOOOONH2

CAS RN 1013921-36-2

IUPAC Name: 3,6,9,12-tetraoxapentadec-14-yn-1-amine
Synonyms: 3,6,9,12-tetraoxapentadec-14-yn-1-amine
Molecular Weight (g/mol): 231.29
Molecular Formula: C11H21NO4
InChi Key: QDLPAHLHHBCWOW-UHFFFAOYSA-N
SMILES: NCCOCCOCCOCCOCC#C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results