CAS RN 10139-84-1
IUPAC Name:
1-(2,4-dihydroxy-3-methylphenyl)ethan-1-one
Synonyms:
1-(2,4-dihydroxy-3-methylphenyl)ethanone
Molecular Weight (g/mol):
166.18
Molecular Formula:
C9H10O3
InChi Key:
KMTLZBUHQPQFAV-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC=C(O)C(C)=C1O
1
–
1
of
1
results
1
Promotions Available
2',4'-Dihydroxy-3'-methylacetophenone, 98%
2',4'-Dihydroxy-3'-methylacetophenone, 98%
CAS: 10139-84-1 Molecular Formula: C9H10O3 Molecular Weight (g/mol): 166.176 MDL Number: MFCD00010817 InChI Key: KMTLZBUHQPQFAV-UHFFFAOYSA-N Synonym: 1-2,4-dihydroxy-3-methylphenyl ethanone,2',4'-dihydroxy-3'-methylacetophenone,1-2,4-dihydroxy-3-methylphenyl ethan-1-one,2,4-dihydroxy-3-methylacetophenone,ethanone, 1-2,4-dihydroxy-3-methylphenyl,acmc-20aocy,1-acetyl-2,4-dihydroxy-3-methylbenzene,2 ',4 '-dihydroxy-3 '-methylacetophenone,1-2,4-dihydroxy-3-methyl-phenyl ethanone,1-2,4-dihydroxy-3-methyl-phenyl-ethanone PubChem CID: 592139 IUPAC Name: 1-(2,4-dihydroxy-3-methylphenyl)ethanone SMILES: CC1=C(C=CC(=C1O)C(=O)C)O