CAS RN 10134-98-2
CAS RN 10134-98-2
IUPAC Name:
1-benzothiophene-7-carboxylic acid
Synonyms:
1-benzothiophene-7-carboxylic acid
Molecular Weight (g/mol):
178.21
Molecular Formula:
C9H6O2S
InChi Key:
LJPSRTWIAXVPIS-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C2SC=CC2=CC=C1
1
–
2
of
2
results
2
1-Benzothiophene-7-carboxylic acid, 97%, Thermo Scientific™
CAS: 10134-98-2 Molecular Formula: C9H6O2S Molecular Weight (g/mol): 178.205 InChI Key: LJPSRTWIAXVPIS-UHFFFAOYSA-N Synonym: benzo b thiophene-7-carboxylic acid,benzo b thiophene-7-carboxylicacid,7-carboxybenzo b thiophene,1-benzo b thiophene-7-carboxylic acid PubChem CID: 14235073 IUPAC Name: 1-benzothiophene-7-carboxylic acid SMILES: C1=CC2=C(C(=C1)C(=O)O)SC=C2