accessibility menu, dialog, popup

UserName

CAS RN 1011-54-7

IUPAC Name: (2E)-3-(2-methoxyphenyl)prop-2-enoic acid
Synonyms: (2E)-3-(2-methoxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: FEGVSPGUHMGGBO-VOTSOKGWSA-N
SMILES: COC1=CC=CC=C1\C=C\C(O)=O
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)