accessibility menu, dialog, popup

UserName

H2OH2OCaOSOOO

CAS RN 10101-41-4

IUPAC Name: calcium dihydrate sulfate
Synonyms: CaSO4.2H2O calcium(II) sulfate, dihydrate (1:1:2) sulfuric acid, calcium salt (1:1), dihydrate sulfuric acid, calcium(2+) salt, dihydrate
Molecular Weight (g/mol): 172.16
Molecular Formula: CaH4O6S
InChi Key: PASHVRUKOFIRIK-UHFFFAOYSA-L
SMILES: O.O.[Ca++].[O-]S([O-])(=O)=O
Quantity 
  • (6)
  • (2)
  • (4)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (9)
  • (2)
Molecular Weight (g/mol) 
  • (34)
  • (2)
Grade 
  • (2)
  • (5)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (4)
  • (3)
  • (1)
Percent Purity 
  • (7)
  • (4)
  • (5)
  • (5)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (11)
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (15)
  • (15)
  • (2)
  • (1)
  • (3)
  • (3)
  • (15)
  • (13)
  • (6)
  • (2)
  • (4)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (9)
  • (2)
  • (34)
  • (2)
  • (2)
  • (5)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (4)
  • (3)
  • (1)
  • (7)
  • (4)
  • (5)
  • (5)
  • (1)
  • (2)
  • (1)
  • (3)
  • (11)
  • (8)