accessibility menu, dialog, popup

UserName

CAS RN 101-80-4

IUPAC Name: 4-(4-aminophenoxy)aniline
Synonyms: 4,4-oxydianiline 4,4'-diaminobiphenyloxide p,p'-diaminodiphenyl ether 4,4-diaminodiphenyl ether 4,4'-oxydiphenylamine 4,4'-diaminobiphenyl oxide 4,4'-diaminodiphenyl oxide ccris 491 dadpe 4,4'-diaminodiphenyl ether
Molecular Weight (g/mol): 200.24
Molecular Formula: C12H12N2O
InChi Key: HLBLWEWZXPIGSM-UHFFFAOYSA-N
SMILES: NC1=CC=C(OC2=CC=C(N)C=C2)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (4)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (1)
  • (5)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)