missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-aminophenyl) ether, 98%

Catalog No. AAA1681536
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1681536 500 g
AAA1681518 50 g
AAA1681522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1681536 Supplier Thermo Scientific Chemicals Supplier No. A1681536

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 101-80-4 | C12H12N2O | 200.241 g/mol

Bis(4-aminophenyl) ether is a monomer used as a cross-linking agent. It is also used in the preparation of poly(imideurea)s, polyimide silica composite membrane, which finds application in gas separation techniques. It is utilized in products such as wire enamels, coatings, adhesives, insulating varnishes, coated fabrics, flame-retardant fibers, oil sealants and retainers. Further, it is used in the preparation of polymer films for electrolyte membranes. In addition to this, it is used as an insulation for cables and printed circuits, and laminates and composite for aerospace vehicles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(4-aminophenyl) ether is a monomer used as a cross-linking agent. It is also used in the preparation of poly(imideurea)s, polyimide silica composite membrane, which finds application in gas separation techniques. It is utilized in products such as wire enamels, coatings, adhesives, insulating varnishes, coated fabrics, flame-retardant fibers, oil sealants and retainers. Further, it is used in the preparation of polymer films for electrolyte membranes. In addition to this, it is used as an insulation for cables and printed circuits, and laminates and composite for aerospace vehicles.

Solubility
Soluble in acetone. Insoluble in water, benzene, carbon tetrachloride, ethanol and hot methanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 101-80-4
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.241
MDL Number MFCD00007863
InChI Key HLBLWEWZXPIGSM-UHFFFAOYSA-N
Synonym 4,4'-oxydianiline, 4,4'-diaminodiphenyl ether, 4,4-oxydianiline, 4-aminophenyl ether, 4,4'-oxybisbenzenamine, oxydianiline, 4-4-aminophenoxy aniline, diaminodiphenyl ether, benzenamine, 4,4'-oxybis, p,p'-oxydianiline
PubChem CID 7579
ChEBI CHEBI:34384
IUPAC Name 4-(4-aminophenoxy)aniline
SMILES C1=CC(=CC=C1N)OC2=CC=C(C=C2)N

Specifications

Melting Point 189°C to 192°C
Boiling Point 190°C (0.1 mmHg)
Flash Point 218°C (424°F)
Quantity 500 g
UN Number UN3077
Beilstein 475735
Solubility Information Soluble in acetone. Insoluble in water,benzene,carbon tetrachloride,ethanol and hot methanol.
Formula Weight 200.24
Percent Purity 98%
Chemical Name or Material Bis(4-aminophenyl) ether
Hazard Category H301+H311+H331-H340-H350-H360
Hazard Statement GHS H Statement
H301-H311-H331-H340-H350-H361
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause genetic defects.
May cause cancer.
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 202-977-0
RTECSNumber BY7900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only