accessibility menu, dialog, popup

UserName

BrO

CAS RN 101-55-3

IUPAC Name: 1-bromo-4-phenoxybenzene
Synonyms: 4-bromophenyl phenyl ether
Molecular Weight (g/mol): 249.11
Molecular Formula: C12H9BrO
InChi Key: JDUYPUMQALQRCN-UHFFFAOYSA-N
SMILES: BrC1=CC=C(OC2=CC=CC=C2)C=C1
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)