missing translation for 'onlineSavingsMsg'
Learn More

4-Bromodiphenyl Ether 98.0+%, TCI America™

Catalog No. B0637500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0637500G 500 g
B063725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0637500G Supplier TCI America Supplier No. B0637500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101-55-3
Molecular Formula C12H9BrO
Molecular Weight (g/mol) 249.107
MDL Number MFCD00000094
InChI Key JDUYPUMQALQRCN-UHFFFAOYSA-N
Synonym 4-bromodiphenyl ether, 4-bromophenyl phenyl ether, 4-bromophenoxybenzene, p-bromodiphenyl ether, p-phenoxybromobenzene, benzene, 1-bromo-4-phenoxy, p-bromophenyl phenyl ether, 4-bromodiphenylether, p-bromophenoxybenzene, ether, p-bromophenyl phenyl
PubChem CID 7565
ChEBI CHEBI:77421
IUPAC Name 1-bromo-4-phenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)Br

Specifications

Color Yellow
Boiling Point 305°C
Quantity 500 g
UN Number 3082
Formula Weight 249.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromodiphenyl Ether
EINECSNumber (3)-0061
TSCA Yes