missing translation for 'onlineSavingsMsg'
Learn More

4-Bromodiphenyl ether, 99%

Catalog No. AAL0147322
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0147322 100 g
AAL0147314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0147322 Supplier Thermo Scientific Chemicals Supplier No. L0147322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 101-55-3 | C12H9BrO | 249.107 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 101-55-3
Molecular Formula C12H9BrO
Molecular Weight (g/mol) 249.107
MDL Number MFCD00000094
InChI Key JDUYPUMQALQRCN-UHFFFAOYSA-N
Synonym 4-bromodiphenyl ether, 4-bromophenyl phenyl ether, 4-bromophenoxybenzene, p-bromodiphenyl ether, p-phenoxybromobenzene, benzene, 1-bromo-4-phenoxy, p-bromophenyl phenyl ether, 4-bromodiphenylether, p-bromophenoxybenzene, ether, p-bromophenyl phenyl
PubChem CID 7565
ChEBI CHEBI:77421
IUPAC Name 1-bromo-4-phenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)Br

Specifications

Melting Point 17°C to 18°C
Density 1.42
Boiling Point 304°C to 305°C
Flash Point >110°C (230°F)
Refractive Index 1.608
Quantity 100 g
UN Number UN3082
Beilstein 1308972
Formula Weight 249.11
Percent Purity 99%
Chemical Name or Material 4-Bromodiphenyl ether
Show More Show Less
Hazard Category H302-H317-H318
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 202-952-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only