CAS RN 10049-16-8
CAS RN 10049-16-8
IUPAC Name:
vanadium pentafluoride
Synonyms:
vanadium pentafluoride
Molecular Weight (g/mol):
145.93
Molecular Formula:
F5V
InChi Key:
LYTSFXQJUMZNFC-UHFFFAOYSA-N
SMILES:
[F].[F].[F].[F].[F].[V]
1
–
1
of
1
results
1
Vanadium(IV) fluoride, 95%
CAS: 10049-16-8 MDL Number: MFCD00042554 Synonym: vanadium iv fluoride,acmc-20aja2,vanadium 4+ tetrafluoride,vanadium 4+ ion tetrafluoride,fluoride, fluoride, fluoride, vanadium salt, fluoride