accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OH2OH2OH2OH2OOSOOOOSOOOOSOOOYbYb

CAS RN 10034-98-7

IUPAC Name: diytterbium(3+) octahydrate trisulfate
Synonyms: diytterbium(3+) octahydrate trisulfate
Molecular Weight (g/mol): 778.4
Molecular Formula: H16O20S3Yb2
InChi Key: MDOSIOHMOWGVIM-UHFFFAOYSA-H
SMILES: O.O.O.O.O.O.O.O.[Yb+3].[Yb+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results