CAS RN 10031-24-0
IUPAC Name:
λ²-tin(2+) dibromide
Synonyms:
tin(II) bromide
Molecular Weight (g/mol):
278.52
Molecular Formula:
Br2Sn
InChi Key:
ZSUXOVNWDZTCFN-UHFFFAOYSA-L
SMILES:
[Br-].[Br-].[Sn++]
1
–
1
of
1
results
1
Tin(II) bromide, 99.2%
CAS: 10031-24-0 Molecular Formula: SnBr2 MDL Number: MFCD00011239 Synonym: tin ii bromide,tin dibromide,stannous bromide,tin bromide snbr2,snbr2,acmc-1bp22