accessibility menu, dialog, popup

UserName

CAS RN 10025-73-7

IUPAC Name: chromium(3+) hexahydrate trichloride
Synonyms: chromium(3+) trichloride hexahydrate
Molecular Weight (g/mol): 266.44
Molecular Formula: Cl3CrH12O6
InChi Key: LJAOOBNHPFKCDR-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
Physical Form 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
Grade 
  • (7)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (8)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (7)
  • (2)
  • (3)