missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Cr3+ Standard for ICP 1000 ppm in 5% HNO3, Ricca Chemical

Catalog No. RV010647100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010647100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV010647100 Supplier Ricca Chemical Company Supplier No. RV010647100N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 5% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

Packaging Natural LDPE Bottle
CAS 10025-73-7, 7697-37-2, 7732-18-5
CAS Min % 92.35%, 4.90%
CAS Max % 96.11%, 5.10%
Specific Gravity 1
Physical Form Liquid
Molecular Formula Cl3CrH12O6
UN Number UN3264
Color Colorless
pH <2
Melting Point Approximately 0°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
InChI Key LJAOOBNHPFKCDR-UHFFFAOYSA-K
Type Standard
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
IUPAC Name chromium(3+) hexahydrate trichloride
Molecular Weight (g/mol) 266.44
Quantity 100 mL
Grade High Purity
Chemical Name or Material Chromium ICP Standard
Show More Show Less