accessibility menu, dialog, popup

UserName

CAS RN 100-80-1

IUPAC Name: 1-ethenyl-3-methylbenzene
Synonyms: M-methylstyrene
Molecular Weight (g/mol): 118.18
Molecular Formula: C9H10
InChi Key: JZHGRUMIRATHIU-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C=C)=C1
Boiling Point 
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)