missing translation for 'onlineSavingsMsg'
Learn More

3-Methylstyrene, 98%, stab. with 0.1% 4-tert-butylcatechol

Catalog No. AAL0807206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0807206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0807206 Supplier Thermo Scientific Chemicals Supplier No. L0807206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 100-80-1 | C9H10 | 118.179 g/mol

3-Methylstyreneis used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylstyreneis used as pharmaceutical intermediates.

Solubility
Soluble in Methanol, Ether, Benzene, Acetone, Ethanol, Heptane. Soluble in water (0.09 g/L) at 20°C.

Notes
Store at 4°C. Pr

Chemical Identifiers

CAS 100-80-1
Molecular Formula C9H10
Molecular Weight (g/mol) 118.179
MDL Number MFCD00008617
InChI Key JZHGRUMIRATHIU-UHFFFAOYSA-N
Synonym 3-methylstyrene, 3-vinyltoluene, 1-methyl-3-vinylbenzene, m-vinyltoluene, m-methylstyrene, benzene, 1-ethenyl-3-methyl, styrene, m-methyl, m-methyl styrene, meta-methylstyrene, unii-ffk4i9430g
PubChem CID 7529
IUPAC Name 1-ethenyl-3-methylbenzene
SMILES CC1=CC=CC(=C1)C=C

Specifications

Melting Point -82°C to -81°C
Density 0.9
Boiling Point 170°C to 171°C
Flash Point 51°C (123°F)
Odor Pungent
Refractive Index 1.541
Quantity 5 g
UN Number UN2618
Beilstein 1304618
Solubility Information Soluble in Methanol,Ether,Benzene,Acetone,Ethanol,Heptane. Soluble in water (0.09g/L) at 20°C.
Formula Weight 118.18
Percent Purity 98%
Chemical Name or Material 3-Methylstyrene, stabilized with 0.1% 4-tert-butylcatechol
Show More Show Less
Hazard Category H226-H304-H315-H319-H332-H335
Hazard Statement GHS H Statement
H304-H226-H315-H319-H335
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: VINYLTOLUENES, STABILIZED
EINECSNumber 202-889-2
RTECSNumber WL5075800
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only