accessibility menu, dialog, popup

UserName

CAS RN 100-25-4

IUPAC Name: 1,4-dinitrobenzene
Synonyms: P-dinitrobenzene
Molecular Weight (g/mol): 168.11
Molecular Formula: C6H4N2O4
InChi Key: FYFDQJRXFWGIBS-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (5)
  • (2)
  • (2)