missing translation for 'onlineSavingsMsg'
Learn More

Motic Swift Line Zoom Stereo Microscope

The M30TZ is ideal for laboratory, industrial and educational use. This stereo microscope features zoom objectives with a continuous magnification range of 7.5X to 40X, offering added versatility that is not an option with fixed power stereo microscopes.

Supplier:  Motic Swift Line M30TZSM99CLBTI1

Encompass

 View more versions of this product

Catalog No. S01438

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • Paired widefield eyepieces are 45° inclined, with diopter adjustment. Interpupillary adjustment from 55 to 75 mm.
  • Trinocular Port accepts video adapter.
  • Interlocked eyepiece tubes with diopter for increased focus acuity.
  • Viewing head rotates 360°. Locks in place by tightening thumb screw.
  • Paired objectives are parfocalled, parcentered, and achromatic.
  • Magnification changes by rotating magnification dial.
  • 3W LED illumination provides bright, white light with virtually no heat.
  • Top light adjusts for optimal lighting angles.
  • Variable transbase illumination allows user to control brightness.
  • Easy-to-replace LED illumination is energy-efficient and rarely requires bulb replacement.
  • Dual illumination switches allow for 3 light settings.
  • Rack and pinion focusing with slip clutch and tension adjustment.
  • Slip-clutch system prevents users from damaging the focusing mechanism.
  • 100 mm reversible black/white plastic contrast plate and 100 mm frosted glass stage plate.
  • Locked-on stage clips.
  • Rugged, cast metal frame.
  • Heavy-duty post mount with locking support collar, permits up or down adjustment of viewing head to accommodate different sized objects on stage.
  • Microscopes with 8 in. tablet or HD Camera available.
Specifications

Chemical Identifiers

7.5X to 40X
Trinocular
WF 10X
Achromatic
LED

Specifications

Zoom Stereo Microscope w/ 8 in. Tablet
Achromatic
LED
7.5X to 40X
Trinocular
WF 10X
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions