missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zolmitriptan 98.0+%, TCI America™
$165.90 - $579.71
Chemical Identifiers
| CAS | 139264-17-8 |
|---|---|
| Molecular Formula | C16H21N3O2 |
| Molecular Weight (g/mol) | 287.36 |
| MDL Number | MFCD00871503 |
| InChI Key | ULSDMUVEXKOYBU-CYBMUJFWSA-N |
| Synonym | zolmitriptan, zomig, zomigoro, zomigon, flezol, zomig-zmt, zomig nasal spray, zomig zmt, zolmitriptane, zolmitriptanum |
| PubChem CID | 60857 |
| ChEBI | CHEBI:10124 |
| IUPAC Name | (4R)-4-({3-[2-(dimethylamino)ethyl]-1H-indol-5-yl}methyl)-1,3-oxazolidin-2-one |
| SMILES | CN(C)CCC1=CNC2=CC=C(C[C@@H]3COC(=O)N3)C=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
Z0024200MG
|
TCI America
Z0024200MG |
200 mg |
Each for $165.90
|
|
|||||
|
Z00241G
|
TCI America
Z00241G |
1 g |
Each for $579.71
|
|
|||||
Chemical Identifiers
| 139264-17-8 | |
| 287.36 | |
| ULSDMUVEXKOYBU-CYBMUJFWSA-N | |
| 60857 | |
| (4R)-4-({3-[2-(dimethylamino)ethyl]-1H-indol-5-yl}methyl)-1,3-oxazolidin-2-one |
| C16H21N3O2 | |
| MFCD00871503 | |
| zolmitriptan, zomig, zomigoro, zomigon, flezol, zomig-zmt, zomig nasal spray, zomig zmt, zolmitriptane, zolmitriptanum | |
| CHEBI:10124 | |
| CN(C)CCC1=CNC2=CC=C(C[C@@H]3COC(=O)N3)C=C12 |
Specifications
| 139264-17-8 | |
| White-Yellow | |
| MFCD00871503 | |
| zolmitriptan, zomig, zomigoro, zomigon, flezol, zomig-zmt, zomig nasal spray, zomig zmt, zolmitriptane, zolmitriptanum | |
| CN(C)CCC1=CNC2=CC=C(C[C@@H]3COC(=O)N3)C=C12 | |
| 287.36 | |
| CHEBI:10124 | |
| ≥98.0% (HPLC,T) | |
| Zolmitriptan |
| 138°C | |
| C16H21N3O2 | |
| 200 mg | |
| ULSDMUVEXKOYBU-CYBMUJFWSA-N | |
| (4R)-4-({3-[2-(dimethylamino)ethyl]-1H-indol-5-yl}methyl)-1,3-oxazolidin-2-one | |
| 60857 | |
| 287.36 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : RQ2707000
TSCA : No
Recommended Storage : Refrigerator