missing translation for 'onlineSavingsMsg'
Learn More

Zolmitriptan 98.0+%, TCI America™

Supplier:  TCI America Z0024200MG

 View more versions of this product

Catalog No. Z0024200MG

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Zolmitriptan
138°C
C16H21N3O2
200 mg
ULSDMUVEXKOYBU-CYBMUJFWSA-N
(4R)-4-({3-[2-(dimethylamino)ethyl]-1H-indol-5-yl}methyl)-1,3-oxazolidin-2-one
60857
287.36
Crystalline Powder
139264-17-8
White-Yellow
MFCD00871503
zolmitriptan, zomig, zomigoro, zomigon, flezol, zomig-zmt, zomig nasal spray, zomig zmt, zolmitriptane, zolmitriptanum
CN(C)CCC1=CNC2=CC=C(C[C@@H]3COC(=O)N3)C=C12
287.36
CHEBI:10124
≥98.0% (HPLC,T)

Chemical Identifiers

139264-17-8
287.36
ULSDMUVEXKOYBU-CYBMUJFWSA-N
60857
(4R)-4-({3-[2-(dimethylamino)ethyl]-1H-indol-5-yl}methyl)-1,3-oxazolidin-2-one
C16H21N3O2
MFCD00871503
zolmitriptan, zomig, zomigoro, zomigon, flezol, zomig-zmt, zomig nasal spray, zomig zmt, zolmitriptane, zolmitriptanum
CHEBI:10124
CN(C)CCC1=CNC2=CC=C(C[C@@H]3COC(=O)N3)C=C12
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : RQ2707000

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions