missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Zolmitriptan | |
| 138°C | |
| C16H21N3O2 | |
| 1 g | |
| ULSDMUVEXKOYBU-CYBMUJFWSA-N | |
| (4R)-4-({3-[2-(dimethylamino)ethyl]-1H-indol-5-yl}methyl)-1,3-oxazolidin-2-one | |
| 60857 | |
| 287.36 | |
| Crystalline Powder |
| 139264-17-8 | |
| White-Yellow | |
| MFCD00871503 | |
| zolmitriptan, zomig, zomigoro, zomigon, flezol, zomig-zmt, zomig nasal spray, zomig zmt, zolmitriptane, zolmitriptanum | |
| CN(C)CCC1=CNC2=CC=C(C[C@@H]3COC(=O)N3)C=C12 | |
| 287.36 | |
| CHEBI:10124 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 139264-17-8 | |
| 287.36 | |
| ULSDMUVEXKOYBU-CYBMUJFWSA-N | |
| 60857 | |
| (4R)-4-({3-[2-(dimethylamino)ethyl]-1H-indol-5-yl}methyl)-1,3-oxazolidin-2-one |
| C16H21N3O2 | |
| MFCD00871503 | |
| zolmitriptan, zomig, zomigoro, zomigon, flezol, zomig-zmt, zomig nasal spray, zomig zmt, zolmitriptane, zolmitriptanum | |
| CHEBI:10124 | |
| CN(C)CCC1=CNC2=CC=C(C[C@@H]3COC(=O)N3)C=C12 |
Safety and Handling
RTECSNumber : RQ2707000
TSCA : No
Recommended Storage : Refrigerator