missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Zoledronic acid monohydrate

Catalog No. AC466210050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466210050 5 g
AC466210010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466210050 Supplier Thermo Scientific Chemicals Supplier No. 466210050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 165800-06-6 | C5H12N2O8P2, C5 H10 N2 O7 P2 . H2 O | 290.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 165800-06-6
Molecular Formula C5 H10 N2 O7 P2 . H2 O

Specifications

Quantity 5 g
Formula Weight 290.11
Physical Form Crystalline Powder
Product Type Reagent
Purity 97.5 to 102.5% (dry substance) (Titration with NaOH)
Chemical Name or Material Zoledronic acid monohydrate

RUO – Research Use Only