missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Zoledronic acid monohydrate

Catalog No. AC466210010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466210010 1 g
AC466210050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466210010 Supplier Thermo Scientific Chemicals Supplier No. 466210010

CAS: 165800-06-6 | C5H12N2O8P2, C5 H10 N2 O7 P2 . H2 O | 290.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 165800-06-6
Molecular Formula C5H12N2O8P2

Specifications

Quantity 1 g
Formula Weight 290.11
Physical Form Crystalline Powder
Product Type Reagent
Purity 97.5 to 102.5% (dry substance) (Titration with NaOH)
InChI Key FUXFIVRTGHOMSO-UHFFFAOYSA-N
SMILES O.OC(CN1C=CN=C1)(P(O)(O)=O)P(O)(O)=O
IUPAC Name [1-hydroxy-2-(1H-imidazol-1-yl)-1-phosphonoethyl]phosphonic acid hydrate
Molecular Weight (g/mol) 290.11
Chemical Name or Material Zoledronic acid monohydrate

RUO – Research Use Only