missing translation for 'onlineSavingsMsg'
Learn More

Zirconium standard solution, for AAS, 1mg/mL Zr in 5% HF, Thermo Scientific Chemicals

$46.85 - $46.85

Specifications

Chemical Name or Material Zirconium standard solution
Name Note 1 mg/ml Zr in 5% HF
CAS 7440-67-7 , 7664-39-3 , 7732-18-5
Physical Form Liquid
Molecular Formula Zr
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC196471000
View Documents
Thermo Scientific Chemicals
196471000
100 mL
Each for $46.85
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for atomic adsorption spectroscopy
Specifications

Specifications

Zirconium standard solution
7440-67-7 , 7664-39-3 , 7732-18-5
Zr
MFCD00011303 MFCD00148878
QCWXUUIWCKQGHC-UHFFFAOYSA-N
zirconium
23995
AAS
1 mg/ml Zr in 5% HF
Liquid
990 to 1010 μg/mL of Zr(4+) in 5% HF
Colorless
[Zr]
91.22
CHEBI:33342
Videos
SDS
Documents

Documents

RUO – Research Use Only