missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ZIPtide Substrate

Catalog No. PV5094
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV5094 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV5094 Supplier Thermo Scientific™ Supplier No. PV5094

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A peptide substrate for the Ca/Calmodulin-dependent protein kinase (CaM) family including CAMK1 and DAPK1.

A peptide substrate for the Ca/Calmodulin-dependent protein kinase (CaM) family including CAMK1 and DAPK1. For use with the Adapta™ Universal Kinase Assay (PV5099).

Specifications

Amino Acid Sequence KKLNRTLSFAEPGW
Assay Entry Biochemical ADP accumulation
Content And Storage 2 mM ZIPtide Peptide substrate in 50 mM HEPES (pH 7.5), 0.01% Brij-35, 10 mM MgCl2 and 1 mM EGTA. Store (at -20°C.)
Detection Method Fluorescence
Quantity 1 mL
Substrate peptide for CaM kinases
Substrate Properties Peptide-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Serine/Threonine Kinases
Gene ID (Entrez) 1612, 8536
Molecular Weight (g/mol) 1646.9 Da
Shipping Condition Dry Ice
Target Entry DAPK1, CAMK1
Validated Kinases CAMK1, DAPK1
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.