missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfate, Certified, LabChem™

Catalog No. p-5388275
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC272701 500 mL
LC272702 1 L
LC272704 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC272701 Supplier LabChem Supplier No. LC272701

Chemical Identifiers

CAS 7446-20-0 , 7732-18-5
Molecular Formula H14O11SZn
Molecular Weight (g/mol) 287.54
MDL Number MFCD00149894
InChI Key RZLVQBNCHSJZPX-UHFFFAOYSA-L
Synonym zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
PubChem CID 62640
ChEBI CHEBI:32312
IUPAC Name zinc(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

Specifications

Color Colorless
CAS Max % 71.5, 28.5
Physical Form Liquid
Quantity 500 mL
Linear Formula Zn(NO3)2·6H2O
Solubility Information Soluble in water
Formula Weight 297.48
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.180g/mL ±0.003
Decomposition Information Sulfur compounds
Density 1.18g/mL
Chemical Name or Material Zinc Sulfate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature