missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfate, Certified, LabChem™

Supplier:  LabChem LC272702

 View more versions of this product

Catalog No. LC272702

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Zinc Sulfate
Colorless
Liquid
H14O11SZn
MFCD00149894
Soluble in water
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O
287.54
CHEBI:32312
Certified
Poly Bottle
Sulfur compounds
7446-20-0,7732-18-5
71.5,28.5
1 L
Zn(NO3)2·6H2O
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
RZLVQBNCHSJZPX-UHFFFAOYSA-L
zinc(2+) heptahydrate sulfate
62640
297.48
Passes Test
1.180g/mL ±0.003
1.18g/mL

Chemical Identifiers

7446-20-0
287.54
RZLVQBNCHSJZPX-UHFFFAOYSA-L
62640
zinc(2+) heptahydrate sulfate
H14O11SZn
MFCD00149894
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
CHEBI:32312
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life.

GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.

Warning

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions