missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfate, 9.5% with Ammonium Hydroxide, Reagents

Catalog No. 01333344
Change view
Click to view available options
:
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01333344 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01333344 Supplier Labchem Inc Supplier No. CZ1322001A
Only null left

Zinc Sulfate, 9.5% with Ammonium Hydroxide, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7446-20-0 , 1336-21-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Zinc Sulphate
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
pH >10
Physical Form Liquid
Quantity 1 L
Percent Purity 9.5%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.07
Chemical Name or Material Zinc Sulfate, 9.5% with Ammonium Hydroxide