missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Sulfate, 0.0500 M (M/20), Ricca Chemical
Specifications
Specifications
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 0.0500M (M/20) |
| Packaging | Natural Poly Bottle |
| pH | ∼4.5 |
| Quantity | 500 mL |
| Chemical Name or Material | Zinc Sulfate |