missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfate, 0.0500 M Aqueous Solution, USP Volumetric Solution, Spectrum™ Chemical

Catalog No. p-7166769
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614803 500 mL Poly Bottle
18614804 1 L Poly Bottle
18614805 4 L Poly Bottle
18614806 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18614803 Supplier Spectrum Chemical Mfg Cor Supplier No. Z351500MLPL
Only null left

Chemical Identifiers

CAS 7733-02-0 , 7732-18-5 , 7446-20-0
Molecular Formula O4SZn
Molecular Weight (g/mol) 161.44
InChI Key NWONKYPBYAMBJT-UHFFFAOYSA-L
IUPAC Name zinc(2+) sulfate
SMILES [Zn++].[O-]S([O-])(=O)=O

Specifications

Quantity 500 mL
Concentration or Composition (by Analyte or Components) 0.050 M ± 0.0005 M
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.