missing translation for 'onlineSavingsMsg'
Learn More

Zinc phosphide, 99.9% (metals basis)

Useful applications in photovoltaic cells. It is also employed as a laboratory chemical. | CAS: 1314-84-7 | P2Zn3

Supplier:  Thermo Scientific Chemicals 03566409

 View more versions of this product

Catalog No. AA3566409


Only null left
Explore available promotions

Description

Description

Zinc phosphide has useful applications in photovoltaic cells. It is also employed as a laboratory chemical. It plays an important role in medicine and pharmaceuticals. It acts as a fumigant in grain storage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1100°C
420°C
Agglomerated Powder
P2Zn3
MFCD00049631
Insoluble in water and ethanol. Soluble in benzene.
99.9%
Moisture sensitive
4.55
Zinc phosphide
1314-84-7
10 g
Zn3P2
UN1714
258.1
Glass bottle
4.55

Chemical Identifiers

1314-84-7
P2Zn3
MFCD00049631
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P223-P231+P232-P264b-P270-P280-P301+P310-P330-P335+P334-P370+P378q-P501c

H260-H300

DOTInformation : Hazard Class: 4.3; Packaging Group: I

EINECSNumber : 215-244-5

RTECSNumber : ZH4900000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only