missing translation for 'onlineSavingsMsg'
Learn More

Zinc phosphide, 99.9% (metals basis)

Catalog No. p-7076347
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
0.5 g
10 g
2 g
Catalog No. Quantity
AA3566402 0.5 g
AA3566404 2 g
AA3566409 10 g
3 options

Catalog No. AA3566402

Supplier: Thermo Scientific Chemicals 03566402

Only null left

Useful applications in photovoltaic cells. It is also employed as a laboratory chemical. | CAS: 1314-84-7 | P2Zn3

Zinc phosphide has useful applications in photovoltaic cells. It is also employed as a laboratory chemical. It plays an important role in medicine and pharmaceuticals. It acts as a fumigant in grain storage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1314-84-7
Molecular Formula P2Zn3
MDL Number MFCD00049631

Specifications

Boiling Point 1100°C
Melting Point 420°C
Physical Form Agglomerated Powder
Quantity 0.5 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Insoluble in water and ethanol. Soluble in benzene.
Formula Weight 258.1
Sensitivity Moisture sensitive
Density 4.55
Chemical Name or Material Zinc phosphide
Hazard Category H260-H300
Precautionary Statement P223-P231+P232-P264b-P270-P280-P301+P310-P330-P335+P334-P370+P378q-P501c
DOTInformation Hazard Class: 4.3; Packaging Group: I
EINECSNumber 215-244-5
RTECSNumber ZH4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only