accessibility menu, dialog, popup

UserName

ZnPHZnPHZn

CAS RN 1314-84-7

IUPAC Name: zinc phosphide
Synonyms: zinc phosphide
Molecular Weight (g/mol): 260.1
Molecular Formula: H2P2Zn3
InChi Key: HOKBIQDJCNTWST-UHFFFAOYSA-N
SMILES: [Zn].P=[Zn].P=[Zn]
Physical Form 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)