missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc(II) Bis(trifluoromethanesulfinate) Dihydrate 98.0+%, TCI America™
Supplier: TCI America Z00285G
Specifications
| Zinc(II) Bis(trifluoromethanesulfinate) Dihydrate | |
| 157°C | |
| C2F6O4S2Zn | |
| 5 g | |
| UANOWFITUWBPCF-UHFFFAOYSA-L | |
| zinc;trifluoromethanesulfinate | |
| 14174735 | |
| ≥98.0% (T) |
| 39971-65-8 | |
| White | |
| MFCD22374174 | |
| Bis(trifluoromethanesulfinic Acid) Zinc(II) Salt, TFMS | |
| C(F)(F)(F)S(=O)[O-].C(F)(F)(F)S(=O)[O-].[Zn+2] | |
| 331.508 | |
| 331.51 | |
| Crystalline Powder |
Chemical Identifiers
| 39971-65-8 | |
| 331.508 | |
| UANOWFITUWBPCF-UHFFFAOYSA-L | |
| 14174735 | |
| C(F)(F)(F)S(=O)[O-].C(F)(F)(F)S(=O)[O-].[Zn+2] |
| C2F6O4S2Zn | |
| MFCD22374174 | |
| Bis(trifluoromethanesulfinic Acid) Zinc(II) Salt, TFMS | |
| zinc;trifluoromethanesulfinate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator