missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc(II) Bis(trifluoromethanesulfinate) Dihydrate 98.0+%, TCI America™
$147.25 - $520.08
Chemical Identifiers
| CAS | 39971-65-8 |
|---|---|
| Molecular Formula | C2F6O4S2Zn |
| Molecular Weight (g/mol) | 331.508 |
| MDL Number | MFCD22374174 |
| InChI Key | UANOWFITUWBPCF-UHFFFAOYSA-L |
| Synonym | Bis(trifluoromethanesulfinic Acid) Zinc(II) Salt, TFMS |
| PubChem CID | 14174735 |
| IUPAC Name | zinc;trifluoromethanesulfinate |
| SMILES | C(F)(F)(F)S(=O)[O-].C(F)(F)(F)S(=O)[O-].[Zn+2] |
Chemical Identifiers
| 39971-65-8 | |
| 331.508 | |
| UANOWFITUWBPCF-UHFFFAOYSA-L | |
| 14174735 | |
| C(F)(F)(F)S(=O)[O-].C(F)(F)(F)S(=O)[O-].[Zn+2] |
| C2F6O4S2Zn | |
| MFCD22374174 | |
| Bis(trifluoromethanesulfinic Acid) Zinc(II) Salt, TFMS | |
| zinc;trifluoromethanesulfinate |
Specifications
| 39971-65-8 | |
| White | |
| MFCD22374174 | |
| Bis(trifluoromethanesulfinic Acid) Zinc(II) Salt, TFMS | |
| C(F)(F)(F)S(=O)[O-].C(F)(F)(F)S(=O)[O-].[Zn+2] | |
| 331.508 | |
| 331.51 | |
| Crystalline Powder |
| 157°C | |
| C2F6O4S2Zn | |
| 1 g | |
| UANOWFITUWBPCF-UHFFFAOYSA-L | |
| zinc;trifluoromethanesulfinate | |
| 14174735 | |
| ≥98.0% (T) | |
| Zinc(II) Bis(trifluoromethanesulfinate) Dihydrate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator